Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-5-fluoronicotinamide
2,6-Dichloro-5-fluoronicotinamide 98%
2,6-Dichloro-5-fluoronicotinamide 98%
Irritant
2,6-Dichloro-5-fluoronicotinamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dichloro-5-fluoronicotinamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
113237-20-0
:
Formula:
C₆H₃Cl₂FN₂O
MDL Number:
MFCD01571365
:
209.01 g/mol
Melting Pt:
161…164 °C
:
GHS07
Storage Temperature:
Ambient