Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluoro-4-nitrophenol
2,5-Difluoro-4-nitrophenol
2,5-Difluoro-4-nitrophenol
Corrosive
Irritant
2,5-Difluoro-4-nitrophenol
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
2,5-Difluoro-4-nitrophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
120103-18-6
:
Formula:
C₆H₃F₂NO₃
MDL Number:
MFCD04039297
:
175.0897 g/mol
:
GHS05
:
GHS07
Storage Temperature:
Ambient