
4-Fluoro-2-methylphenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:139911-29-8
:MFCD17214439
Formula:C₇H₈BFO₂
MDL Number:MFCD02093072
:153.95 g/mol
Melting Pt:191…196 °C
:GHS07
Storage Temperature:Refrigerator