Order Entry
Export
ContactUsLinkComponent
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Catalog # APOSPC6673-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Catalog # APOSPC6673-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :241.01 g/mol
Storage Temperature:Ambient
CAS Number:79456-34-1
Formula:C₆H₄BrF₃N₂
MDL Number:MFCD10758088