Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,5-difluorobenzoic acid
2-Bromo-4,5-difluorobenzoic acid 98%
2-Bromo-4,5-difluorobenzoic acid 98%
Irritant
2-Bromo-4,5-difluorobenzoic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Bromo-4,5-difluorobenzoic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
64695-84-7
:
Formula:
C₇H₃BrF₂O₂
MDL Number:
MFCD04973422
:
237.0000 g/mol
Melting Pt:
118…122 °C
:
GHS07
Storage Temperature:
Ambient