Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized
Fomblin® Z-60
Fomblin® Z-60
Irritant
Fomblin® Z-60
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Fomblin® Z-60
Supplier:
Apollo Scientific
About this item
CAS Number:
69991-67-9
:
MDL Number:
MFCD00163699
Boiling Pt:
>270 °C
Melting Pt:
–63 °C
Density:
1.85 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Ambient