Order Entry
Export
Orders LinkContactUsLinkComponent
Poly(vinylidene fluoride)

Poly(vinylidene fluoride)

Irritant
Supplier:   Apollo Scientific
Poly(vinylidene fluoride)
Supplier:  Apollo Scientific
Certificates
Poly(vinylidene fluoride)

3 Options Available Below

Product Details & Documents
  • CAS Number:24937-79-9
  • Formula:(-CH₂CF₂-)n
  • MDL Number:MFCD00084470
  • Melting Pt:155…165 °C
  • Density:1.74 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 57,90 €
    57,90 €
    1 * 100 g
       57,90 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 123,00 €
    123,00 €
    1 * 250 g
       123,00 €