

1,1,1-Trifluoro-6-methyl-2,4-heptanedione
Catalog # APOSPC6237-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.17 g/mol
Boiling Pt:78 °C/64mmHg
Density:1.13 g/cm³ (25 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:207-319-6
UN:1224
ADR:3,III
:GHS02
:GHS07
CAS Number:461-92-7
Formula:C₈H₁₁F₃O₂
MDL Number:MFCD00045107