Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
APOSPC6152-500MG
:
Apollo Scientific
:
4,5-Dibromo-1-methyl-1H-imidazole-2-carbaldehyde
APOSPC6152-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
267.92 g/mol
:
167…169 °C
:
GHS07
:
Ambient
:
79326-92-4
:
C₅H₄Br₂N₂O
:
MFCD12025924