
1-Iodoperfluorohexane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:355-43-1
:
Formula:C₆F₁₃I
MDL Number:MFCD00001063
:445.95 g/mol
Boiling Pt:115…117 °C
Melting Pt:–45 °C
Density:2.063 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:206-586-6
:GHS07