Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2,3-difluoropyridine
5-Bromo-2,3-difluoropyridine 98%
5-Bromo-2,3-difluoropyridine 98%
Corrosive
Irritant
5-Bromo-2,3-difluoropyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
5-Bromo-2,3-difluoropyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
89402-44-8
:
Formula:
C₅H₂BrF₂N
MDL Number:
MFCD06659525
:
193.98 g/mol
UN:
3265
ADR:
8,III
:
GHS05
:
GHS07
Storage Temperature:
Ambient