Order Entry
Export
ContactUsLinkComponent
2,5-Bis(trifluoromethyl)phenylacetonitrile 98%
Toxic
2,5-Bis(trifluoromethyl)phenylacetonitrile 98%
Catalog # APOSPC5971-1G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
2,5-Bis(trifluoromethyl)phenylacetonitrile 98%
Catalog # APOSPC5971-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
 :GHS06
CAS Number:302911-99-5
Formula:C₁₀H₅F₆N