Order Entry
Export
ContactUsLinkComponent
2-(3-Bromo-4-(trifluoromethoxy)phenyl)-2,2-difluoroacetic acid 95%
2-(3-Bromo-4-(trifluoromethoxy)phenyl)-2,2-difluoroacetic acid 95%
Catalog # APOSPC5754-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Bromo-4-(trifluoromethoxy)phenyl)-2,2-difluoroacetic acid 95%
Catalog # APOSPC5754-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Refrigerator
CAS Number:1133116-05-8
Formula:C₉H₄BrF₅O₃
MDL Number:MFCD11855929