Methyl 5-bromo-3-fluoropicolinate 95%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Methyl 5-bromo-3-fluoropicolinate, Methyl 5-bromo-3-fluoropyridine-2-carboxylate
- CAS Number:1211538-72-5
- Formula:C₇H₅BrFNO₂
- MDL Number:MFCD14698121
- Molecular Weight:234.03 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Methyl 5-bromo-3-fluoropicolinate, Methyl 5-bromo-3-fluoropyridine-2-carboxylate
- CAS Number:1211538-72-5
- Formula:C₇H₅BrFNO₂
- MDL Number:MFCD14698121
- Molecular Weight:234.03 g/mol
- Storage Temperature:Ambient



