Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethyl-4-fluoroaniline
2,6-Dimethyl-4-fluoroaniline 98%
2,6-Dimethyl-4-fluoroaniline 98%
Irritant
2,6-Dimethyl-4-fluoroaniline 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dimethyl-4-fluoroaniline 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
392-70-1
:
Formula:
C₈H₁₀FN
MDL Number:
MFCD03792669
:
139.17 g/mol
:
GHS07
Storage Temperature:
Refrigerator