
(Pentafluorophenyl)diphenylphosphine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5525-95-1
:
Formula:C₁₈H₁₀F₅P
MDL Number:MFCD00000290
:352.24 g/mol
Melting Pt:70…72 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient