Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Poly(chlorotrifluoroethylene) 800
Oil 10 S
Oil 10 S
:
Apollo Scientific
Oil 10 S
:
Apollo Scientific
:
9002-83-9
:
:
(C₂ClF₃)
n
:
800 g/mol
:
–129 °C
:
1.71 g/cm³ (25 °C)
:
GHS07
:
Ambient