

1,1,1,4,4,4-Hexafluoro-2,3-butanedione
Catalog # APOSPC56260-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:2.1,
:194.03 g/mol
UN:3161
:GHS07
:GHS04
Storage Temperature:Ambient
CAS Number:685-24-5
Formula:C₄F₆O₂