

1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887144-97-0
:
Formula:C₁₀H₁₀F₃IO
MDL Number:MFCD10567056
:330.09 g/mol
Melting Pt:75…79 °C
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Storage Temperature:Refrigerator