0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4,5-Difluoro-o-toluidine
4,5-Difluoro-o-toluidine 95%
4,5-Difluoro-o-toluidine 95%
Irritant
4,5-Difluoro-o-toluidine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,5-Difluoro-o-toluidine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
875664-57-6
:
Formula:
C₇H₇F₂N
MDL Number:
MFCD08062378
:
143.14 g/mol
Melting Pt:
56…58 °C
:
GHS07
Storage Temperature:
Ambient