Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluoro-4-iodopyridine
2,5-Difluoro-4-iodopyridine
2,5-Difluoro-4-iodopyridine
:
Apollo Scientific
2,5-Difluoro-4-iodopyridine
:
Apollo Scientific
:
1017793-20-2
:
:
C₅H₂F₂IN
:
MFCD11040606
:
240.98 g/mol
:
209.7 °C
:
80…85 °C
:
GHS07
:
Ambient