
3,3,3-Trifluoro-2,2-dimethylpropionic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:889940-13-0
:
Formula:C₅H₇F₃O₂
MDL Number:MFCD08445819
:156.11 g/mol
Boiling Pt:76…77 °C
Melting Pt:66…71 °C
:GHS07
Storage Temperature:Ambient