0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4,4-Difluoropiperidine hydrochloride
4,4-Difluoropiperidine hydrochloride
4,4-Difluoropiperidine hydrochloride
Irritant
4,4-Difluoropiperidine hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,4-Difluoropiperidine hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
144230-52-4
:
Formula:
C₅H₁₀ClF₂N
MDL Number:
MFCD03095381
:
157.59 g/mol
Melting Pt:
173…177 °C
:
GHS07
Storage Temperature:
Refrigerator