Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,6-difluorobenzoic acid
4-Chloro-2,6-difluorobenzoic acid 98%
4-Chloro-2,6-difluorobenzoic acid 98%
Irritant
4-Chloro-2,6-difluorobenzoic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Chloro-2,6-difluorobenzoic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
196194-58-8
:
Formula:
C₇H₃ClF₂O₂
MDL Number:
MFCD06203570
:
192.55 g/mol
Melting Pt:
182…186 °C
:
GHS07
Storage Temperature:
Ambient