
α,α,α-Trifluoro-2,3-diaminotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:360-60-1
:
Formula:C₇H₇F₃N₂
MDL Number:MFCD08532495
:176.14
Boiling Pt:125 °C/14mmHg
Melting Pt:41 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07