Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Bis(trifluoromethyl)-2-chloropyridine
3,6-Bis(trifluoromethyl)-2-chloropyridine 98%
3,6-Bis(trifluoromethyl)-2-chloropyridine 98%
APOSPC520889-1G
:
Apollo Scientific
:
3,6-Bis(trifluoromethyl)-2-chloropyridine 98%
APOSPC520889-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
249.54 g/mol
:
Ambient
:
175136-26-2
:
C₇H₂ClF₆N
:
MFCD00128907