Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3,6-difluorobenzotrifluoride
2-Chloro-3,6-difluorobenzotrifluoride 95%
2-Chloro-3,6-difluorobenzotrifluoride 95%
APOSPC520794-5G
:
Apollo Scientific
:
2-Chloro-3,6-difluorobenzotrifluoride 95%
APOSPC520794-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
216.54 g/mol
:
Ambient
:
1099597-70-2
:
C₇H₂ClF₅
:
MFCD11226658