Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Difluoromethyl)-2,6-difluoro-pyridine
3-(Difluoromethyl)-2,6-difluoro-pyridine 97%
3-(Difluoromethyl)-2,6-difluoro-pyridine 97%
:
Apollo Scientific
3-(Difluoromethyl)-2,6-difluoro-pyridine 97%
:
Apollo Scientific
:
85396-63-0
:
:
C₆H₃F₄N
:
MFCD22199312
:
165.09 g/mol
:
GHS07
:
Ambient