Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-4-(1,1-difluoropropyl)benzene
1-Bromo-4-(1,1-difluoropropyl)benzene 95%
1-Bromo-4-(1,1-difluoropropyl)benzene 95%
1-Bromo-4-(1,1-difluoropropyl)benzene 95%
APOSPC520724-1G
:
Apollo Scientific
:
1-Bromo-4-(1,1-difluoropropyl)benzene 95%
APOSPC520724-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
235.08 g/mol
:
Ambient
:
162783-79-1
:
C₉H₉BrF₂
:
MFCD16140148