Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-6-(1,1-difluoroethyl)pyridine
2-Bromo-6-(1,1-difluoroethyl)pyridine 95%
2-Bromo-6-(1,1-difluoroethyl)pyridine 95%
2-Bromo-6-(1,1-difluoroethyl)pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-6-(1,1-difluoroethyl)pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1211535-69-1
:
Formula:
C₇H₆BrF₂N
MDL Number:
MFCD16140181
:
222.04 g/mol
Storage Temperature:
Ambient