Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(1,1-Difluoroethyl)-3-nitrobenzene
1-(1,1-Difluoroethyl)-3-nitrobenzene 95%
1-(1,1-Difluoroethyl)-3-nitrobenzene 95%
1-(1,1-Difluoroethyl)-3-nitrobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(1,1-Difluoroethyl)-3-nitrobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
40788-04-3
:
Formula:
C₈H₇F₂NO₂
MDL Number:
MFCD14525560
:
187.15 g/mol
Storage Temperature:
Ambient