Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-Chloro-1,1-difluoroethyl)benzene
(2-Chloro-1,1-difluoroethyl)benzene 95%
(2-Chloro-1,1-difluoroethyl)benzene 95%
(2-Chloro-1,1-difluoroethyl)benzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
(2-Chloro-1,1-difluoroethyl)benzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
55805-08-8
:
Formula:
C₈H₇ClF₂
MDL Number:
MFCD14525555
:
176.59 g/mol
Storage Temperature:
Ambient