Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(1,1-Difluoroethyl)pyridine
3-(1,1-Difluoroethyl)pyridine 97%
3-(1,1-Difluoroethyl)pyridine 97%
Toxic
3-(1,1-Difluoroethyl)pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
3-(1,1-Difluoroethyl)pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
114468-03-0
:
Formula:
C₇H₇F₂N
MDL Number:
MFCD12404951
:
143.14 g/mol
Storage Temperature:
Ambient
UN:
2811
ADR:
6.1,III
:
GHS06