Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(1,1-Difluoroethyl)pyrazine
2-(1,1-Difluoroethyl)pyrazine 95%
2-(1,1-Difluoroethyl)pyrazine 95%
2-(1,1-Difluoroethyl)pyrazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(1,1-Difluoroethyl)pyrazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
111781-49-8
:
Formula:
C₆H₆F₂N₂
MDL Number:
MFCD12546932
:
144.12 g/mol
Storage Temperature:
Ambient