Order Entry
Export
ContactUsLinkComponent
1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate 90%
1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate 90%
Catalog # APOSPC520675-25G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(2,4-Difluorophenyl)-2,3-epoxypropyl]-1H-1,2,4-triazole methanesulphonate 90%
Catalog # APOSPC520675-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :333.31 g/mol
Storage Temperature:Ambient
CAS Number:86386-77-8
 :MFCD13184268
Formula:C₁₂H₁₃F₂N₃O₄S
MDL Number:MFCD11111978