Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%
Irritant
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,2-Dichloro-4,5-bis(trifluoromethyl)benzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
320-22-9
:
Formula:
C₈H₂Cl₂F₆
MDL Number:
MFCD11100166
:
283 g/mol
:
GHS07
Storage Temperature:
Ambient