Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-2-(trifluoromethyl)pyridine
3,5-Dichloro-2-(trifluoromethyl)pyridine 97%
3,5-Dichloro-2-(trifluoromethyl)pyridine 97%
Toxic
3,5-Dichloro-2-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
3,5-Dichloro-2-(trifluoromethyl)pyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
7655-72-3
:
Formula:
C₆H₂Cl₂F₃N
MDL Number:
MFCD11226518
:
215.99 g/mol
Storage Temperature:
Ambient
UN:
2810
ADR:
6.1,III
:
GHS06