Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-3-nitrobenzotrifluoride
2-Methyl-3-nitrobenzotrifluoride 97%
2-Methyl-3-nitrobenzotrifluoride 97%
:
Apollo Scientific
2-Methyl-3-nitrobenzotrifluoride 97%
:
Apollo Scientific
:
6656-49-1
:
:
C₈H₆F₃NO₂
:
MFCD00042322
:
205.14 g/mol
:
86 °C/10mmHg
:
>100 °C
:
Ambient
:
229-688-2
:
2810
:
6.1,III
:
GHS06