Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Difluoro-m-toluidine
2,6-Difluoro-m-toluidine 98%
2,6-Difluoro-m-toluidine 98%
Irritant
2,6-Difluoro-m-toluidine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Difluoro-m-toluidine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
144851-63-8
:
Formula:
C₇H₇F₂N
MDL Number:
MFCD06660182
:
143.14 g/mol
Storage Temperature:
Refrigerator
UN:
2810
ADR:
6.1,III
:
GHS07