Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
APOSPC51033-1G
:
Apollo Scientific
:
6-Fluoroimidazo[1,2-a]pyridine-2-carbohydrazide
APOSPC51033-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
263…264 °C
:
194.17 g/mol
:
GHS07
:
Ambient
:
1135282-88-0
:
C₈H₇FN₄O
:
MFCD11840960