
4,6-Dichloro-5-fluoropyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:213265-83-9
:
Formula:C₄HCl₂FN₂
MDL Number:MFCD08056331
:166.97 g/mol
Boiling Pt:50…60 °C/14mmHg
Flash Pt:91 °C
Storage Temperature:Ambient
UN:1760
ADR:8,II
:GHS05