Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3,4-difluorobenzoate
Methyl-3,4-difluorobenzoate 97%
Methyl-3,4-difluorobenzoate 97%
:
Apollo Scientific
Methyl-3,4-difluorobenzoate 97%
:
Apollo Scientific
:
369-25-5
:
:
C₈H₆F₂O₂
:
MFCD00276597
:
172.13 g/mol
:
87…89 °C/15mmHg
:
23…27 °C
:
GHS05
:
GHS07
:
Ambient