Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromo-α,α,α-trifluorotoluene
3,5-Dibromo-α,α,α-trifluorotoluene 96%
3,5-Dibromo-α,α,α-trifluorotoluene 96%
Irritant
3,5-Dibromo-α,α,α-trifluorotoluene 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dibromo-α,α,α-trifluorotoluene 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
401-84-3
:
Formula:
C₇H₃Br₂F₃
MDL Number:
MFCD00526219
:
303.90 g/mol
:
GHS07
Storage Temperature:
Ambient