Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,2-difluoro-2-(fluorosulphonyl)acetate
Methyl-2,2-difluoro-2-(fluorosulphonyl)acetate 97%
Methyl-2,2-difluoro-2-(fluorosulphonyl)acetate 97%
:
Apollo Scientific
Methyl-2,2-difluoro-2-(fluorosulphonyl)acetate 97%
:
Apollo Scientific
:
680-15-9
:
:
C₃H₃F₃O₄S
:
MFCD00144316
:
117…118 °C
:
1.509 g/cm³ (25 °C)
:
47 °C
:
2920
:
8+3,II
:
192.11 g/mol
:
GHS02
:
GHS05
:
Ambient