Order Entry
Export
Orders LinkContactUsLinkComponent
Manganese(II) fluoride 99%

Manganese(II) fluoride 99%

Bottle
Catalog # APOSPC5030-500G
Irritant
New Product
Supplier:   Apollo Scientific
Manganese(II) fluoride 99%
Catalog # APOSPC5030-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  7782-64-1
Manganese(II) fluoride 99%

Bottle

CAS Number:  7782-64-1
Price
78,20 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Manganese(II) difluoride, Manganese difluoride
  • CAS Number:7782-64-1
  • Formula:F₂Mn
  • MDL Number:MFCD00016222
  • Molecular Weight:92.93 g/mol
  • Melting Pt:856 °C
  • Density:3.98 g/cm³ (25 °C)
  •  :Density: 3.98 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications