Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichloro-7-fluoroquinazoline
2,4-Dichloro-7-fluoroquinazoline 95%
2,4-Dichloro-7-fluoroquinazoline 95%
2,4-Dichloro-7-fluoroquinazoline 95%
APOSPC502107-1G
:
Apollo Scientific
:
2,4-Dichloro-7-fluoroquinazoline 95%
APOSPC502107-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
217.0272 g/mol
:
Ambient
:
174566-15-5
:
C₈H₃Cl₂FN₂
:
MFCD09954885