Potassium-1,3-dimethyluracil-5-trifluoroborate
Catalog # APOSPC50058-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Refrigerator
CAS Number:1150654-77-5
Formula:C₆H₇BF₃KN₂O₂
MDL Number:MFCD09271782