Order Entry
Export
ContactUsLinkComponent
 
3,5-Dichloro-4-fluorobenzaldehyde
Catalog # APOSPC500504-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dichloro-4-fluorobenzaldehyde
Catalog # APOSPC500504-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.0000 g/mol
Storage Temperature:Ambient
CAS Number:117820-80-1
Formula:C₇H₃Cl₂FO
MDL Number:MFCD11845864