Order Entry
Export
ContactUsLinkComponent
1-(Difluoromethyl)-2,4-difluorobenzene
1-(Difluoromethyl)-2,4-difluorobenzene
Catalog # APOSPC49631-5G
Supplier:  Apollo Scientific
CAS Number:  
1-(Difluoromethyl)-2,4-difluorobenzene
Catalog # APOSPC49631-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.1 g/mol
Storage Temperature:Ambient
CAS Number:845866-84-4
Formula:C₇H₄F₄
MDL Number:MFCD06657968