Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dimethoxytetrafluorobenzene
1,4-Dimethoxytetrafluorobenzene
1,4-Dimethoxytetrafluorobenzene
1,4-Dimethoxytetrafluorobenzene
APOSPC4955-5G
:
Apollo Scientific
:
1,4-Dimethoxytetrafluorobenzene
APOSPC4955-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
51.1…52.5 °C
:
GHS07
:
Ambient
:
362-56-1
:
C₈H₆F₄O₂